Naphthalene-1,4-dicarboxylic acid, 98+%, Thermo Scientific Chemicals
Naphthalene-1,4-dicarboxylic acid, 98+%, Thermo Scientific Chemicals
Naphthalene-1,4-dicarboxylic acid, 98+%, Thermo Scientific Chemicals
Naphthalene-1,4-dicarboxylic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Naphthalene-1,4-dicarboxylic acid, 98+%, Thermo Scientific Chemicals
CAS: 605-70-9 | C12H8O4 | 216.19 g/mol
Have Questions?
Catalog NumberQuantity
ALFL20249.1425 g
Catalog number ALFL20249.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS508-02-1
IUPAC Name(4aS,6aS,6bR,8aR,10S,12aR,12bR,14bS)-10-hydroxy-2,2,6a,6b,9,9,12a-heptamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-4a-carboxylic acid
Molecular FormulaC30H48O3
InChI KeyMIJYXULNPSFWEK-GTOFXWBISA-N
SMILESCC1(C)CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]2C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
Water=<5 %
Appearance (Form)Powder
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Tapas Kumar Maji; Wakako Kaneko; Masaaki Ohba; and Susumu Kitagawa. Diversity in magnetic properties of 3D isomorphous networks of Co(II) and Mn(II) constructed by napthalene-1,4-dicarboxylate. Chem. Commun. 2005, (36), 4613-4615.
  2. J. F. Walker; N. D. Scott. Sodium Naphthalene. II. Preparation and Properties of Dihydronaphthalene Dicarboxylic Acids. J. Am. Chem. Soc. 1938, (60), 951-955.