1-Ethyl-3-methylimidazolium bromide, 98+%, Thermo Scientific Chemicals
1-Ethyl-3-methylimidazolium bromide, 98+%, Thermo Scientific Chemicals
1-Ethyl-3-methylimidazolium bromide, 98+%, Thermo Scientific Chemicals
1-Ethyl-3-methylimidazolium bromide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Ethyl-3-methylimidazolium bromide, 98+%, Thermo Scientific Chemicals
CAS: 65039-08-9 | C6H11BrN2 | 191.07 g/mol
Have Questions?
Catalog NumberQuantity
ALFL19761.1850 g
Catalog number ALFL19761.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS1724-39-6
IUPAC Namecyclododecanol
Molecular FormulaC12H24O
InChI KeySFVWPXMPRCIVOK-UHFFFAOYSA-N
SMILESOC1CCCCCCCCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or fused/lumpy solid
Appearance (Color)White
Assay (GC)≥98.5%
Melting Point (clear melt)75.0-81.0?C
An ionic liquid, 1-ethyl-3-methylimidazolium bromide is used as reaction medium for the synthesis and crystallization of a coordination polymer, (EMI)[Cd(BTC)]. 1-ethyl-3-methylimidazolium bromide was used as a solvent in [emim] 2 [Cd 2 (btec)Br 2 ]. Form the diene complexation; carbonyl insertion; cycloaddition; reduction and deoxygenation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
An ionic liquid, 1-ethyl-3-methylimidazolium bromide is used as reaction medium for the synthesis and crystallization of a coordination polymer, (EMI)[Cd(BTC)]. 1-ethyl-3-methylimidazolium bromide was used as a solvent in [emim] 2 [Cd 2 (btec)Br 2 ]. Form the diene complexation; carbonyl insertion; cycloaddition; reduction and deoxygenation.
Solubility
Soluble in water, dichloromethane, ethanol, acetonitrile. Insoluble in ethyl acetate, ether, and alkane.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
RUO – Research Use Only
General References:
  1. Mohammed Taghi Zafarani-Moattar,; Shokat Sarmad. Effect of tri-potassium phosphate on volumetric, acoustic, and transport behaviour of aqueous solutions of 1-ethyl-3-methylimidazolium bromide at T = (298.15 to 318.15) K. The Journal of Chemical Thermodynamics. 2010, 42 (10), 1213-1221.
  2. Steven D. Chambreau,; Jerry A. Boatz,; Ghanshyam L. Vaghjiani,; Christine Koh,; Oleg Kostko.Thermal Decomposition Mechanism of 1-Ethyl-3-methylimidazolium Bromide Ionic Liquid. J. Phys. Chem. A. 2012 , 116 (24), 5867-5876.