Amberlite XAD-16, Thermo Scientific Chemicals
Amberlite XAD-16, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Amberlite XAD-16, Thermo Scientific Chemicals
CAS: 104219-63-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFL19565.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS66-84-2
IUPAC Namehydrogen 2-amino-3,4,5,6-tetrahydroxyhexanal chloride
Molecular FormulaC6H14ClNO5
InChI KeyCBOJBBMQJBVCMW-UHFFFAOYNA-N
SMILES[H+].[Cl-].NC(C=O)C(O)C(O)C(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
Optical Rotation72.5 ± 1.5? (C=2 in water, 24hrs)
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
Amberlite XAD-16 is a polymeric adsorbent widely utilized for adsorption of organic substances from aqueous systems and polar solvents. It is useful for the removal of detergents from protein solutions. It is also used in the recovery and purification of antibiotics, water soluble steroids, amino acids and proteins. It plays a vital role in the removal of non polar compounds from polar solvents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Amberlite XAD-16 is a polymeric adsorbent widely utilized for adsorption of organic substances from aqueous systems and polar solvents. It is useful for the removal of detergents from protein solutions. It is also used in the recovery and purification of antibiotics, water soluble steroids, amino acids and proteins. It plays a vital role in the removal of non polar compounds from polar solvents.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents and acids.
RUO – Research Use Only
General References:
  1. Non-ionic polystyrene adsorbent, approximate surface area 800 m 2 /g.
  2. Amin, A. S.; Optimized solid phase extraction methodology for separation, and spectrophotometric determination of boron using amberlite XAD-16 resin modified with 2-(2-benzothiazolylazo)-4-methoxyphenol. RSC Adv. 2015, 5 (82), 66975-66980.
  3. Islam, A.; Akil, A.; Asaduddin, L. M.; Flame Atomic Absorption Spectrometric Determination of Trace Metal Ions in Environmental and Biological Samples After Preconcentration on a Newly Developed Amberlite XAD-16 Chelating Resin Containing p-Aminobenzene Sulfonic Acid. J. AOCA Int. 2015, 98 (1), 165-175.