Amberlite XAD-7, Thermo Scientific Chemicals
Amberlite XAD-7, Thermo Scientific Chemicals
Amberlite XAD-7, Thermo Scientific Chemicals
Amberlite XAD-7, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Amberlite XAD-7, Thermo Scientific Chemicals
CAS: 37380-43-1
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFL19564.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmberlite XAD-7
CAS37380-43-1
Recommended StorageAmbient temperatures
TSCANo
MDL NumberMFCD00132705
View more
Amberlite XAD-7 is used in chromatography media, enzyme immobilization and water purification. It is also used in the removal of relatively polar compounds from non aqueous solvents, non-aromatic compounds from polar solvent, recovery of plant extracts, antibiotics, enzymes and proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Amberlite XAD-7 is used in chromatography media, enzyme immobilization and water purification. It is also used in the removal of relatively polar compounds from non aqueous solvents, non-aromatic compounds from polar solvent, recovery of plant extracts, antibiotics, enzymes and proteins.
Solubility
Insoluble in water.
Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Francisco A. Tomás-Barberán.; M. Amparo Blázquez.; Cristina Garcia-Viguera.; Federico Ferreres.; Francisco Tomás-Lorente. A comparative study of different amberlite XAD resins in flavonoid analysis. Phytochemical Analysis. 1992, 3 (4), 178-181.
  2. B Saha.; R.J Gill.; D.G Bailey.; N Kabay.; M Arda. Sorption of Cr(VI) from aqueous solution by Amberlite XAD-7 resin impregnated with Aliquat 336. Reactive and Functional Polymers. 2004, 60 223-244.
  3. Non-ionic polyacrylate adsorbent, approximate surface area 450 m 2 /g.