1,4-Benzenedithiol, 97%, Thermo Scientific Chemicals
1,4-Benzenedithiol, 97%, Thermo Scientific Chemicals
1,4-Benzenedithiol, 97%, Thermo Scientific Chemicals
1,4-Benzenedithiol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Benzenedithiol, 97%, Thermo Scientific Chemicals
CAS: 624-39-5 | C6H6S2 | 142.23 g/mol
Have Questions?
Catalog NumberQuantity
ALFL19440.065 g
Catalog number ALFL19440.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS2052-49-5
IUPAC Nametetrabutylazanium hydroxide
Molecular FormulaC16H37NO
InChI KeyVDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES[OH-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to slightly hazy colorless to light yellow
Appearance (Form)Solution
Titration with HClO4>=40 %
1,4-Benzenedithiol is used in single molecule junctions in electronic devices. It is also used to treat silver nano particles and in the preparation of gold electrodes. It finds an application in nanomaterial synthesis and in self-assembled monolayer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,4-Benzenedithiol is used in single molecule junctions in electronic devices. It is also used to treat silver nano particles and in the preparation of gold electrodes. It finds an application in nanomaterial synthesis and in self-assembled monolayer.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
General References:
  1. Matsuhita, R.; Horikawa, M.; Naitoh, Y.; Nakamura, H.; Kiguchi, M. Conductance and SERS Measurement of Benzenedithiol Molecules Bridging Between Au Electrodes. J. Phys. Chem., C 2013,117 (4), 1791-1795.
  2. Vidal-Iglesias, F. J.; Solla-Gullón, J.; Orts, J. M.; Rodes, A.; Pérez, J. M. Spectroelectrochemical Study of the Photoinduced Catalytic Formation of 4,4'-Dimercaptoazobenzene from 4-Aminobenzenethiol Adsorbed on Nanostructured Copper. J. Phys. Chem. C 2015, 119 (22), 12312-12324.