5-Bromo-2-methyl-2-pentene, 97%, Thermo Scientific Chemicals
5-Bromo-2-methyl-2-pentene, 97%, Thermo Scientific Chemicals
5-Bromo-2-methyl-2-pentene, 97%, Thermo Scientific Chemicals
5-Bromo-2-methyl-2-pentene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Bromo-2-methyl-2-pentene, 97%, Thermo Scientific Chemicals
CAS: 2270-59-9 | C6H11Br | 163.06 g/mol
Have Questions?
Catalog NumberQuantity
ALFL19406.065 g
Catalog number ALFL19406.06
Price (MYR)
1,808.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,808.00
Add to cart
Specifications
Chemical Name or Material5-Bromo-2-methyl-2-pentene
CAS2270-59-9
Health Hazard 1H226-H315-H319-H335
Health Hazard 2GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
View more
5-Bromo-2-methyl-2-pentene acts as a synthetic reagent, which is useful in the formation of sesqueterpenoid. It is also employed in the preparation of geranlol-3-14C. Further, it is used in the preparation of penifulvin A, which is a sesquiterpenoid with a novel dioxa-fenestrane structure.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Bromo-2-methyl-2-pentene acts as a synthetic reagent, which is useful in the formation of sesqueterpenoid. It is also employed in the preparation of geranlol-3-14C. Further, it is used in the preparation of penifulvin A, which is a sesquiterpenoid with a novel dioxa-fenestrane structure.
Solubility
Slightly miscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only
General References:
  1. Wills, V. S.; Allen, C.; Holstein, S. A.; Wiemer, D. F. Potent Triazole Bisphosphonate Inhibitor of Geranylgeranyl Diphosphate Synthase. ACS Med. Chem. Lett. 2015, 6 (12), 1195-1198.
  2. Langhanki, J.; Rudolph, K.; Erkel, G.; Opatz, T. Total synthesis and biological evaluation of the natural product (-)-cyclonerodiol, a new inhibitor of IL-4 signaling. Org. Biomol. Chem. 2014, 12 (47), 9707-9715.