1-n-Butyl-3-methylimidazolium hexafluorophosphate, 98+%, Thermo Scientific Chemicals
1-n-Butyl-3-methylimidazolium hexafluorophosphate, 98+%, Thermo Scientific Chemicals
1-n-Butyl-3-methylimidazolium hexafluorophosphate, 98+%, Thermo Scientific Chemicals
1-n-Butyl-3-methylimidazolium hexafluorophosphate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-n-Butyl-3-methylimidazolium hexafluorophosphate, 98+%, Thermo Scientific Chemicals
CAS: 174501-64-5 | C8H15F6N2P | 284.19 g/mol
Have Questions?
Catalog NumberQuantity
ALFL19086.22100 g
Catalog number ALFL19086.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS7782-49-2
IUPAC Nameselenium
Molecular FormulaSe
InChI KeyBUGBHKTXTAQXES-UHFFFAOYSA-N
SMILES[Se]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark grey
Appearance (Form)Tablets with green or blue speckles
Nitrogen (N)=<0.05 %
Additional infoCopper sulphate pentahydrate (per tablet):
Additional info68 to 82 mg
View more
1-n-Butyl-3-methylimidazolium hexafluorophosphate is used as a solvent in headspace gas chromatography and countercurrent chromatography, as mobile and stationary phases. It is widely used in organic synthesis, extraction technologies, electochemistry and biphasic organometallic catalysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-n-Butyl-3-methylimidazolium hexafluorophosphate is used as a solvent in headspace gas chromatography and countercurrent chromatography, as mobile and stationary phases. It is widely used in organic synthesis, extraction technologies, electochemistry and biphasic organometallic catalysis.
Solubility
Miscible with dichloromethane, chloroform, ethyl acetate. Immiscible with water, diethyl ether and hexane.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Soni, S. K.; Sarkar, S.; Mirzadeh , N.; Selvakannan , P. R.; Bhargava, S. K. Self-Assembled Functional Nanostructure of Plasmid DNA with Ionic Liquid [Bmim][PF6]: Enhanced Efficiency in Bacterial Gene Transformation. Langmuir. 2015, 31 (16), 4722-4732.
  2. Zhang, Y.; Zhang, T.; Gan, P.; Li, H.; Zhang, M.; Jin, K.; Tang, S. Solubility of Isobutane in Ionic Liquids [BMIm][PF6], [BMIm][BF4], and [BMIm][Tf2N]. J. Chem. Eng. Data. 2015, 60 (6), 1706-1714.