5-Chloro-2-(2,4-dichlorophenoxy)phenol, 99%, Thermo Scientific Chemicals
5-Chloro-2-(2,4-dichlorophenoxy)phenol, 99%, Thermo Scientific Chemicals
5-Chloro-2-(2,4-dichlorophenoxy)phenol, 99%, Thermo Scientific Chemicals
5-Chloro-2-(2,4-dichlorophenoxy)phenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Chloro-2-(2,4-dichlorophenoxy)phenol, 99%, Thermo Scientific Chemicals
CAS: 3380-34-5 | C12H7Cl3O2 | 289.536 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
10 g
100 g
Catalog number ALFL18655.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS16718-11-9
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.6500-1.6550 @ 20?C
5-Chloro-2-(2,4-dichlorophenoxy)phenol, acts as a bacteriostat and preservative for cosmetic and detergent compositions. It is also used as an antiseptic, disinfectant and anticonvulsant. It is one of the main ingredient in shampoos, deodorants, toothpastes, mouth washes and household cleaning products. It inhibits the growth of bacteria, fungi on the surfaces of the textile, thereby it imparts odor protection to wool, synthetics, blends and non-wovens.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Chloro-2-(2,4-dichlorophenoxy)phenol, acts as a bacteriostat and preservative for cosmetic and detergent compositions. It is also used as an antiseptic, disinfectant and anticonvulsant. It is one of the main ingredient in shampoos, deodorants, toothpastes, mouth washes and household cleaning products. It inhibits the growth of bacteria, fungi on the surfaces of the textile, thereby it imparts odor protection to wool, synthetics, blends and non-wovens.
Solubility
Soluble in ethanol, methanol, diethyl ether and sodium hydroxide solution (1M). Slightly soluble in water.
Notes
Store in cool place. Decomposes on heating. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Halden, R. U. On the Need and Speed of Regulating Triclosan and Triclocarban in the United States. Environ. Sci. Technol. 2014, 48 (7), 3603-3611.
  2. Gao, Y.; Ji, Y.; Li, G.; An, T. Mechanism, kinetics and toxicity assessment of OH-initiated transformation of triclosan in aquatic environments. Water Res. 2014, 49, 360-370.