3,5-Dihydroxytoluene, 99%, Thermo Scientific Chemicals
3,5-Dihydroxytoluene, 99%, Thermo Scientific Chemicals
3,5-Dihydroxytoluene, 99%, Thermo Scientific Chemicals
3,5-Dihydroxytoluene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,5-Dihydroxytoluene, 99%, Thermo Scientific Chemicals
CAS: 504-15-4 | C7H8O2 | 124.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFL18567.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS401-81-0
IUPAC Name1-iodo-3-(trifluoromethyl)benzene
Molecular FormulaC7H4F3I
InChI KeyIGISPMBUGPHLBY-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CC(I)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Refractive Index1.5165-1.5205 @ 20°C
Appearance (Color)Clear colorless to pale yellow or pale pink
Identification (FTIR)Conforms
FormLiquid
Used in production of orcein