3,5-Dihydroxytoluene, 99%, Thermo Scientific Chemicals
3,5-Dihydroxytoluene, 99%, Thermo Scientific Chemicals
3,5-Dihydroxytoluene, 99%, Thermo Scientific Chemicals
3,5-Dihydroxytoluene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,5-Dihydroxytoluene, 99%, Thermo Scientific Chemicals
CAS: 504-15-4 | C7H8O2 | 124.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFL18567.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS451-46-7
IUPAC Nameethyl 4-fluorobenzoate
Molecular FormulaC9H9FO2
InChI KeyUMPRJGKLMUDRHL-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light yellow
Appearance (Form)Low melting solid
GC>=98.5 %
Infrared spectrumConforms
Refractive index1.4850 to 1.4870 (20°C, 589 nm)
Used in production of orcein