Isobutyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals
Isobutyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals
Isobutyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals
Isobutyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isobutyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals
CAS: 4247-02-3 | C11H14O3 | 194.23 g/mol
Have Questions?
Catalog NumberQuantity
ALFL17587.1850 g
Catalog number ALFL17587.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialIsobutyl 4-hydroxybenzoate
CAS4247-02-3
Melting Point74°C to 78°C
Recommended StorageAmbient temperatures
RTECS NumberDH2247000
View more
Isobutyl 4-hydroxybenzoate is used as an anti-microbial agent. It is also used as antiseptics for cosmetic field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isobutyl 4-hydroxybenzoate is used as an anti-microbial agent. It is also used as antiseptics for cosmetic field.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Zhu, K.; Gong, S.; Dai, Y., Tuo, S.; Liu, Z.; Qian, L.; Zhou, F. Determination of Preservatives in Water-Based Adhesives by Gas Chromatography-Mass Spectrometry. Analytical Letters. 2015, 48 (9), 1396-1410.
  2. Sanchez-Prado, L.; Alvarez-Rivera, G.; Lamas, J. P.; Lores, M.; Garcia-Jares, C.; Llompart, M. Analysis of multi-class preservatives in leave-on and rinse-off cosmetics by matrix solid-phase dispersion. Analytical and bioanalytical chemistry. 2011, 401 (10), 3293-3304..