Sodium perfluorooctanoate, 97%, Thermo Scientific™
Sodium perfluorooctanoate, 97%, Thermo Scientific™
Sodium perfluorooctanoate, 97%, Thermo Scientific™
Sodium perfluorooctanoate, 97%, Thermo Scientific™
Sodium perfluorooctanoate, 97%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFL16988.1425 g
Catalog number ALFL16988.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium perfluorooctanoate
CAS335-95-5
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Melting Point∽275°C (decomposition)
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Sodium perfluorooctanoate on gel-to-liquid-crystalline phase transition of dipalmitoylphosphatidylcholine vesicle membrane.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
General References:
  1. Norbert Muller.; Harvey Simsohn. Investigation of micelle structure by fluorine magnetic resonance. V. Sodium perfluorooctanoate. J. Phys. Chem. 1971, 75 (7), 942-945.
  2. Stuart S. Berr.; Richard R. M. Jones. Small-angle neutron scattering from aqueous solutions of sodium perfluorooctanoate above the critical micelle concentration. J. Phys. Chem. 1989, 93 (6), 2555-2558.