4,4'-(Hexafluoroisopropylidene)diphthalic anhydride, 99%, Thermo Scientific Chemicals
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride, 99%, Thermo Scientific Chemicals
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride, 99%, Thermo Scientific Chemicals
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride, 99%, Thermo Scientific Chemicals
CAS: 1107-00-2 | C19H6F6O6 | 444.241 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Catalog number ALFL16150.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS9006-59-1
MDL NumberMFCD00130424
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Powder
Protein>=80 %
Water=<8.0 % (K.F.)
4,4'-(Hexafluoroisopropylidene) diphthalic anhydride is involved in the synthesis of polyamides derived from terphenylenes, which is used for gas separation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,4′-(Hexafluoroisopropylidene) diphthalic anhydride is involved in the synthesis of polyamides derived from terphenylenes, which is used for gas separation.
Solubility
Miscible with water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
General References:
  1. Raaijmakers, M. J. T.; Kappert, E. J.; Nijmeijer, A.; Benes, N. E. Thermal Imidization Kinetics of Ultrathin Films of Hybrid Poly(POSS-imide)s. Macromolecules 2015, 48 (9), 3031-3039.
  2. Tapaswi, P. K.; Choi, M.; Jeong, K.; Ando, S.; Ha, C. Transparent Aromatic Polyimides Derived from Thiophenyl-Substituted Benzidines with High Refractive Index and Small Birefringence. Macromolecules 2015, 48 (11), 3462-3474.