Di-n-propylamine, 99%, Thermo Scientific Chemicals
Di-n-propylamine, 99%, Thermo Scientific Chemicals
Di-n-propylamine, 99%, Thermo Scientific Chemicals
Di-n-propylamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Di-n-propylamine, 99%, Thermo Scientific Chemicals
CAS: 142-84-7 | C6H15N | 101.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number ALFL15808.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS771-60-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting crystalline solid
Infrared spectrumConforms
Melting point33°C to 38°C
Appearance (Color)White to pink-orange
GC>=96.0 %