(+/-)-Citronellal, 96%, Thermo Scientific Chemicals
(+/-)-Citronellal, 96%, Thermo Scientific Chemicals
(+/-)-Citronellal, 96%, Thermo Scientific Chemicals
(+/-)-Citronellal, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(+/-)-Citronellal, 96%, Thermo Scientific Chemicals
CAS: 106-23-0 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALFL15753.AP
Price (MYR)
881.00
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
881.00
Add to cart
Specifications
Chemical Name or Material(+/-)-Citronellal
CAS106-23-0
Health Hazard 1H227-H315-H317-H319
Health Hazard 2GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Health Hazard 3P210-P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P370+P378q-P501c
View more
(±)-Citronellal is an active component of citronella oil. It is used as a flavoring agent, insect repellent, fragrance in soaps and detergents, and in personal care products like creams, lotions, and perfumes. It acts as a chemical intermediate for citronellol and hydroxycitronellal. Further, it is used to prepare 1-dibutylamino-3,7-dimethyl-oct-6-en-2-one by reacting with dibutylamine. In addition to this, it is utilized to inhibit the development of mechanical nociception induced by carrageenan (CG) and tumor necrosis factor-alfa.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(^+)-Citronellal is an active component of citronella oil. It is used as a flavoring agent, insect repellent, fragrance in soaps and detergents, and in personal care products like creams, lotions, and perfumes. It acts as a chemical intermediate for citronellol and hydroxycitronellal. Further, it is used to prepare 1-dibutylamino-3,7-dimethyl-oct-6-en-2-one by reacting with dibutylamine. In addition to this, it is utilized to inhibit the development of mechanical nociception induced by carrageenan (CG) and tumor necrosis factor-alfa.
Solubility
Slightly miscible with water and ethanol.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Air and light sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only
General References:
  1. Itoh, H.; Maeda, H.; Yamada, S.; Hori, Y.; Mino, T.; Sakamoto, M. BINOL-Al catalysed asymmetric cyclization and amplification: preparation of optically active menthol analogs. Org. Biomol. Chem. 2015, 13 (20), 5817-5825.
  2. George, J.; Shalabney, A.; Hutchison, J. A.; Genet, C.; Ebbesen, T. W. Liquid-Phase Vibrational Strong Coupling. J. Phys. Chem. Lett. 2015, 6 (6), 1027-1031.