4,4'-Methylenebis(phenyl isocyanate), 98%, Thermo Scientific Chemicals
4,4'-Methylenebis(phenyl isocyanate), 98%, Thermo Scientific Chemicals
4,4'-Methylenebis(phenyl isocyanate), 98%, Thermo Scientific Chemicals
4,4'-Methylenebis(phenyl isocyanate), 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4,4'-Methylenebis(phenyl isocyanate), 98%, Thermo Scientific Chemicals
CAS: 101-68-8 | C15H10N2O2 | 250.257 g/mol
Have Questions?
Catalog NumberQuantity
ALFL15740.36500 g
Catalog number ALFL15740.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material4,4'-Methylenebis(phenyl isocyanate)
CAS101-68-8
Health Hazard 1H315-H317-H319-H332-H334-H335-H351-H373
Health Hazard 2GHS H Statement
H330-H334-H351-H373-H315-H319-H317-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3P201-P202-P260-P264b-P271-P272-P280-P281-P285-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P342+P311-P362-P501c
View more
4,4'-Methylenebis(phenyl isocyanate) acts as a monomer in the production of polyurethane. The rigid polyurethane foams are good thermal insulators, which find applications in all freezers and refrigerators, as well as in buildings. It is also used as an industrial strength adhesive, which is available to end consumers as various high-strength bottled glue preparations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,4′-Methylenebis(phenyl isocyanate) acts as a monomer in the production of polyurethane. The rigid polyurethane foams are good thermal insulators, which find applications in all freezers and refrigerators, as well as in buildings. It is also used as an industrial strength adhesive, which is available to end consumers as various high-strength bottled glue preparations.
Solubility
Insoluble in water. Soluble in acetone, benzene and kerosene.
Notes
Incompatible with water, amines, strong bases, alcohols and oxidizing agents.
RUO – Research Use Only
Chong, D. S. T.; Turner, L. A.; Gadegaard, N.; Seifalian, A. M.; Dalby, M. J.; Hamilton, G. Nanotopography and Plasma Treatment: Redesigning the Surface for Vascular Graft Endothelialisation. Eur. J. Vasc. Endovasc. 2015, 49 (3), 335-343.