4,4'-Oxybis(benzoic acid), 98+%, Thermo Scientific Chemicals
4,4'-Oxybis(benzoic acid), 98+%, Thermo Scientific Chemicals
4,4'-Oxybis(benzoic acid), 98+%, Thermo Scientific Chemicals
4,4'-Oxybis(benzoic acid), 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4,4'-Oxybis(benzoic acid), 98+%, Thermo Scientific Chemicals
CAS: 2215-89-6 | C14H10O5 | 258.229 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALFL15651.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS110-94-1
IUPAC Namepentanedioic acid
Molecular FormulaC5H8O4
InChI KeyJFCQEDHGNNZCLN-UHFFFAOYSA-N
SMILESOC(=O)CCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
Titration with NaOH98.5 to 101.5 %
Melting point95°C to 99°C