1-Octadecylamine, 97%, Thermo Scientific Chemicals
1-Octadecylamine, 97%, Thermo Scientific Chemicals
1-Octadecylamine, 97%, Thermo Scientific Chemicals
1-Octadecylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Octadecylamine, 97%, Thermo Scientific Chemicals
CAS: 124-30-1 | C18H39N | 269.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALFL15458.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS156-41-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Refractive index1.5474 to 1.5494 (20°C, 589 nm)
Infrared spectrumConforms
Water=<0.5 % (K.F.)
View more
1-Octadecylamine is used as asphalt emulsifier, lubricant and dispersants. It acts as a cationic surfactant and find applications is hair rinse bases, wood preservatives, textile softeners, dyeing auxiliaries and pigment grinding aids. It is used in boiler as an additive, which prevents the corrosion arises due to steam. It is a basic building block and used as an intermediate in the preparation of their amine derivatives, ethoxylates and amides. Metal oxide nano crystals having controlled size and shape are prepared by the thermal decomposition of metal nitrates with octadecylamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Octadecylamine is used as asphalt emulsifier, lubricant and dispersants. It acts as a cationic surfactant and find applications is hair rinse bases, wood preservatives, textile softeners, dyeing auxiliaries and pigment grinding aids. It is used in boiler as an additive, which prevents the corrosion arises due to steam. It is a basic building block and used as an intermediate in the preparation of their amine derivatives, ethoxylates and amides. Metal oxide nano crystals having controlled size and shape are prepared by the thermal decomposition of metal nitrates with octadecylamine.
Solubility
Soluble in chloroform, alcohol, ether, benzene and acetone.Insoluble in water.
Notes
Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
RUO – Research Use Only
General References:
  1. Wang, S.; Zhang, c.; Li, Y.; Li, B.; Yang, Y. Chirality of Single-Handed Twisted Titania Tubular Nanoribbons Prepared Through Sol-gel Transcription. Chirality 2015, 27 (8), 543-550.
  2. Ng, Y. H.; Tay, S. W.; Hong, R. S.; Hong, L. In situ formation of reverse polymeric micelles in liquid alkanes to lodge alcohol micro-droplets. RSC Adv. 2015, 5 (27), 21033-21041.