Check Order Status
Acetaldehyde, 99%
Structure Search
Thermo Scientific Chemicals
Acetaldehyde, 99%
Have Questions?
Change view
Quantity:
500 mL
Catalog number
ALFL15215.AP
Price (MYR)
646.00
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
646.00
Add to cart
Specifications
Quantity
500 mL
Percent Purity
0.99
Unit Size