Methacrylamide, 98%, Thermo Scientific Chemicals
Methacrylamide, 98%, Thermo Scientific Chemicals
Methacrylamide, 98%, Thermo Scientific Chemicals
Methacrylamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methacrylamide, 98%, Thermo Scientific Chemicals
CAS: 79-39-0 | C4H7NO | 85.106 g/mol
Have Questions?
Catalog NumberQuantity
ALFL15013.22100 g
Catalog number ALFL15013.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS60811-23-6
IUPAC Name(3-chloro-4-fluorophenyl)sulfanide
Molecular FormulaC6H3ClFS
InChI KeySFSHSIFYTWIGSF-UHFFFAOYSA-M
SMILESFC1=CC=C([S-])C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Refractive Index1.5705-1.5755 @ 20?C
Methacrylamide is a reagent used in the production of polymers and copolymers. It is also used as a raw material in the production of chemicals for industrial and professional use such as textiles, leather, fur, bulk, large scale chemicals (including petroleum products), fine chemicals, formulation [mixing] of preparations and/or re-packaging (excluding alloys), building and construction work, electricity, steam, gas, water supply and sewage treatment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methacrylamide is a reagent used in the production of polymers and copolymers. It is also used as a raw material in the production of chemicals for industrial and professional use such as textiles, leather, fur, bulk, large scale chemicals (including petroleum products), fine chemicals, formulation [mixing] of preparations and/or re-packaging (excluding alloys), building and construction work, electricity, steam, gas, water supply and sewage treatment.
Solubility
Soluble in water (202g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Laura M. Y. Yu.; Karineh Kazazian.; Molly S. Shoichet. Peptide surface modification of methacrylamide chitosan for neural tissue engineering applications. Journal of Biomedical Materials Research Part A. 2007, 82A (1), 243-255.
  2. Burcin Akgun.; Emre Savci.; Duygu Avci. Synthesis and polymerizations of phosphonated-bis(methacrylamide)s for dental applications. Journal of Polymer Science Part A: Polymer Chemistry. 2012, 50 (4), 801-810.