Bisphenol A diglycidyl ether resin, Thermo Scientific Chemicals
Bisphenol A diglycidyl ether resin, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bisphenol A diglycidyl ether resin, Thermo Scientific Chemicals
CAS: 1675-54-3 | C21H24O4 | 340.419 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number ALFL14817.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.2% max.
Bisphenol A diglycidyl ether is used to inhibit the peroxisome proliferator-activated receptor (PPARγ) and adipocyte differentiation and it induce apoptosis in tumor cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bisphenol A diglycidyl ether is used to inhibit the peroxisome proliferator-activated receptor (PPARγ) and adipocyte differentiation and it induce apoptosis in tumor cells.
Solubility
Soluble in 100% ethanol, dimethyl sulfoxide (100 mM), dimethyl formamide, chloroform, methanol, and ethanol (50 mM). Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Neus Casajuana.; Sílvia Lacorte. New Methodology for the Determination of Phthalate Esters, Bisphenol A, Bisphenol A Diglycidyl Ether, and Nonylphenol in Commercial Whole Milk Samples . J. Agric. Food Chem. 2004, 52 (12), 3702-3707.
  2. P. Paseiro Losada.; J. Simal Lozano.; S. Paz Abuín.; P. López Mahía.; J. Simal Gándara. Kinetics of the hydrolysis of bisphenol A diglycidyl ether (BADGE) in water-based food simulants. Fresenius' Journal of Analytical Chemistry. 1993, 345 (7), 527-532.