Chloroform, ethanol-free, 99+%, stab. with ca 50 ppm amylene, Thermo Scientific Chemicals
Chloroform, ethanol-free, 99+%, stab. with ca 50 ppm amylene, Thermo Scientific Chemicals
Chloroform, ethanol-free, 99+%, stab. with ca 50 ppm amylene, Thermo Scientific Chemicals
Chloroform, ethanol-free, 99+%, stab. with ca 50 ppm amylene, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroform, ethanol-free, 99+%, stab. with ca 50 ppm amylene, Thermo Scientific Chemicals
CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Have Questions?
Catalog NumberQuantity
ALFL14759.0F2500 mL
Catalog number ALFL14759.0F
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2500 mL
Request bulk or custom format
Chemical Identifiers
CAS7791-18-6
IUPAC Namemagnesium(2+) hexahydrate dichloride
Molecular FormulaCl2H12MgO6
InChI KeyDHRRIBDTHFBPNG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystals or crystalline powder
Assay from Suppliers CofAMagnesium Chloride (MgCl₂): ≥45.4% (≥97.0% purity calculated as hexahydrate)
Impurity contentCalcium: Report %
Impurity contentPotassium: Report %
View more
Chloroform is a commonly used laboratory solvent. It was previously used as an anesthetic. It is also used as a precursor to PTFE and refrigerants. Chloroform was used in the past as an extraction solvent for fats, oils and greases. It is useful for the rapid and simple release of periplasmic proteins. The mixture of guanidinium thiocyanate-phenol-chloroform, plays an essential role in the RNA isolation process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chloroform is a commonly used laboratory solvent. It was previously used as an anesthetic. It is also used as a precursor to PTFE and refrigerants. Chloroform was used in the past as an extraction solvent for fats, oils and greases. It is useful for the rapid and simple release of periplasmic proteins. The mixture of guanidinium thiocyanate-phenol-chloroform, plays an essential role in the RNA isolation process.
Solubility
Miscible with diethyl ether, oils, ligroin, alcohol, acetone, carbon tetrachloride and carbondisulfide.Slightly miscible with water.
Notes
It is a stable and light sensitive chemical compound. Incompatible with metals, alkalies. Keep the material away from heat, sparks and flame and other sources of ignition. Store it in a cool, dry, well-ventilated area..
RUO – Research Use Only
General References:
  1. Fong, J. K.; Pena, J. K.; Xue, Z.; Alam, M. M.; Sampathkumaran, U.; Goswani, K. Optical Sensors for the Detection of Trace Chloroform. Anal. Chem. 2015, 87 (3), 1569-1574.
  2. Yadav, R.; Balasubramanian, K.; Wang, X. Encapsulation of gold nanoparticles with PHB based on coffee ring effect. RSC Adv. 2015, 5 (24), 18501-18505.