Isoprene, 99%, stab. with ca 0.02% 4-tert-butylcatechol, Thermo Scientific Chemicals
Isoprene, 99%, stab. with ca 0.02% 4-tert-butylcatechol, Thermo Scientific Chemicals
Isoprene, 99%, stab. with ca 0.02% 4-tert-butylcatechol, Thermo Scientific Chemicals
Isoprene, 99%, stab. with ca 0.02% 4-tert-butylcatechol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isoprene, 99%, stab. with ca 0.02% 4-tert-butylcatechol, Thermo Scientific Chemicals
CAS: 78-79-5 | C5H8 | 68.119 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number ALFL14619.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS73-40-5
IUPAC Name2-amino-6,7-dihydro-3H-purin-6-one
Molecular FormulaC5H5N5O
InChI KeyUYTPUPDQBNUYGX-UHFFFAOYSA-N
SMILESNC1=NC(=O)C2=C(N1)N=CN2
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<0.5 %
Ratio of Absorbanceslambda max. 248 nm, pH 1
Ratio of Absorbances0.82 to 0.86 (A280/A260)
Heavy metals (as Pb)=<10 ppm
View more
Isoprene is used in to prepare synthetic version of natural rubber by polymerization using Ziegler catalysts. It is a basic building block for terpenes especially carotenes, terpenoids, retinol, vitamin-A and tocopherol (Vitamin-E). It finds applications in medical equipments, toys, baby bottle nipples, shoe soles, elastic films and threads for golf balls. Copolymer prepares in association with styrene is used in pressure sensitive adhesives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isoprene is used in to prepare synthetic version of natural rubber by polymerization using Ziegler catalysts. It is a basic building block for terpenes especially carotenes, terpenoids, retinol, vitamin-A and tocopherol (Vitamin-E). It finds applications in medical equipments, toys, baby bottle nipples, shoe soles, elastic films and threads for golf balls. Copolymer prepares in association with styrene is used in pressure sensitive adhesives.
Solubility
Miscible with ethanol, ethyl ether, alcohol, acetone and benzene. Immiscible with water.
Notes
Store in a cool place. Incompatible with oxidizing agents, strong bases, ammonia, alkali metals, alcohols and oxygen.
RUO – Research Use Only
General References:
  1. Kuwata, M.; Liu, Y.; McKinney, K.; Martin, S. T. Physical state and acidity of inorganic sulfate can regulate the production of secondary organic material from isoprene photooxidation products. Phys. Chem. Chem. Phys. 2015, 17 (8), 5670-5678.
  2. Loreto, F.; Fineschi, S. Reconciling functions and evolution of isoprene emission in higher plants. New Phytol. 2015, 206 (2), 578-582.