Filter Aid, Celite Standard Supercel∣r
Filter Aid, Celite Standard Supercel∣r
Filter Aid, Celite Standard Supercel∣r
Filter Aid, Celite Standard Supercel∣r
Thermo Scientific Chemicals
Filter Aid, Celite Standard Supercel∣r
CAS: 61790-53-2 | O2Si | 60.083 g/mol
Have Questions?
Catalog NumberQuantity
ALFL14552.36500 g
Catalog number ALFL14552.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialFilter Aid, Celite AW Standard Supercel™
OdorOdorless
Quantity500 g
CAS61790-53-2
Health Hazard 1H319-H335-H373
View more
Filter Aid, Celite AW Standard Supercel™ is used for general filtrations. It is also used as a functional filler in paints, plastics, paper, rubber, adhesives, catalysts, agricultural chemicals, pharmaceuticals and other chemicals, and a mild abrasive in polishes and cleaners. It is also used in clarifying liquids and oils, as an efficient thermal insulator; in manufacture of heat insulators, fire brick, and fire- and acid-proof packing materials; as an absorbent; in absorbent dynamite; as a solid carrier in chromatography; pozzolanic admixture in concrete mixes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Filter Aid, Celite AW Standard Supercel« is used for general filtrations. It is also used as a functional filler in paints, plastics, paper, rubber, adhesives, catalysts, agricultural chemicals, pharmaceuticals and other chemicals,
RUO – Research Use Only
General References:
  1. S. E Calvert. Accumulation of Diatomaceous Silica in the Sediments of the Gulf of California. Geological Society of America Bulletin. 1965, 77 (6), 569-596.
  2. Engel G. Vrieling; Theo P. M. Beelen; Rutger A. Van Santen andWinfried W. C. Gieskes. Nanoscale uniformity of pore architecture in diatomaceous silica: a combined small and wide angle x-ray scattering study. Journal of Phycology. 2000, 36 (1), 146-159.
  3. Celite is a registered trademark of Celite Corp.
  4. Acid-washed grade for general filtrations.