D-Glucuronic acid, 98+%, Thermo Scientific Chemicals
D-Glucuronic acid, 98+%, Thermo Scientific Chemicals
D-Glucuronic acid, 98+%, Thermo Scientific Chemicals
D-Glucuronic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
D-Glucuronic acid, 98+%, Thermo Scientific Chemicals
CAS: 6556-12-3 | C6H10O7 | 194.14 g/mol
Have Questions?
Catalog NumberQuantity
ALFL14350.065 g
Catalog number ALFL14350.06
Price (MYR)
445.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
445.00
Add to cart
Specifications
Chemical Name or MaterialD-Glucuronic acid
CAS6556-12-3
Melting Point∽158°C (decomposition)
Recommended StorageAmbient temperatures
RTECS NumberLZ8836600
View more
D-Glucuronic acid is used as pharmaceutical intermediate and in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-Glucuronic acid is used as pharmaceutical intermediate and in chemical research.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. J. J. Burns.; A. H. Conney.; Peter G. Dayton.; Carole Evans.; G. R. Martin.; Dolores Taller. Observations On The Drug-Induced Synthesis Of D-Glucuronic, L-Gulonic And L-Ascorbic Acids In Rats. Jpet. 1960, 129 (2), 132-138.E5
  2. Ulf Lindahl.; Gudrun Bäckström.; Anders Malmström.; Lars-Åke Fransson. Biosynthesis of L-iduronic acid in heparin: Epimerization of D-glucuronic acid on the polymer level. Biochemical and Biophysical Research Communications. 1972, 46 (2), 985-991.