2-Aminoanthracene, 94%, Thermo Scientific Chemicals
2-Aminoanthracene, 94%, Thermo Scientific Chemicals
2-Aminoanthracene, 94%, Thermo Scientific Chemicals
2-Aminoanthracene, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Aminoanthracene, 94%, Thermo Scientific Chemicals
CAS: 613-13-8 | C14H11N | 193.249 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFL14269.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS102-47-6
IUPAC Name1,2-dichloro-4-(chloromethyl)benzene
Molecular FormulaC7H5Cl3
InChI KeyYZIFVWOCPGPNHB-UHFFFAOYSA-N
SMILESClCC1=CC=C(Cl)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.5756 to 1.5776 (20°C, 589 nm)
GC>=98.0 %