Dowex™ 1X2 100-200 (Cl), Thermo Scientific Chemicals
Dowex™ 1X2 100-200 (Cl), Thermo Scientific Chemicals
Dowex™ 1X2 100-200 (Cl), Thermo Scientific Chemicals
Dowex™ 1X2 100-200 (Cl), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dowex™ 1X2 100-200 (Cl), Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFL14254.0B1000 g
Catalog number ALFL14254.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Chemical Identifiers
CAS144-55-8
IUPAC Namesodium hydrogen carbonate
Molecular FormulaCHNaO3
InChI KeyUIIMBOGNXHQVGW-UHFFFAOYSA-M
SMILES[Na+].OC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with HCl99.4 to 100.6 %
SolubilityClear colorless
Heavy metals (as Pb)=<5 ppm
UVat 280 nm A: =<0.02