5,6,7-Trihydroxyflavone, 97%, Thermo Scientific Chemicals
5,6,7-Trihydroxyflavone, 97%, Thermo Scientific Chemicals
5,6,7-Trihydroxyflavone, 97%, Thermo Scientific Chemicals
5,6,7-Trihydroxyflavone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5,6,7-Trihydroxyflavone, 97%, Thermo Scientific Chemicals
CAS: 491-67-8 | C15H10O5 | 270.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
100 mg
Catalog number ALFL14185.ME
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mg
Request bulk or custom format
Chemical Identifiers
CAS14755-02-3
IUPAC Name(2E)-3-(3-hydroxyphenyl)prop-2-enoate
Molecular FormulaC9H7O3
InChI KeyKKSDGJDHHZEWEP-SNAWJCMRSA-M
SMILESOC1=CC=CC(\C=C\C([O-])=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)>98.5%
Appearance (Color)White to cream
Assay (GC)>98.5% (silyl derivative)
Formcrystalline powder