5,6,7-Trihydroxyflavone, 97%, Thermo Scientific Chemicals
5,6,7-Trihydroxyflavone, 97%, Thermo Scientific Chemicals
5,6,7-Trihydroxyflavone, 97%, Thermo Scientific Chemicals
5,6,7-Trihydroxyflavone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5,6,7-Trihydroxyflavone, 97%, Thermo Scientific Chemicals
CAS: 491-67-8 | C15H10O5 | 270.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number ALFL14185.MC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS288-32-4
IUPAC Name1H-imidazole
Molecular FormulaC3H4N2
InChI KeyRAXXELZNTBOGNW-UHFFFAOYSA-N
SMILESN1C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
CommentPurchased in the U.K. and in other countries
Water Content (Karl Fischer Titration)≤0.5%
Appearance (Color)White to pale cream to pale yellow
FormCrystals or flakes or powder or crystalline flakes or crystalline powder
Assay (GC)≥98.5% (U.K. specification)