(S)-(+)-2-Butanol, 98+%, Thermo Scientific Chemicals
(S)-(+)-2-Butanol, 98+%, Thermo Scientific Chemicals
(S)-(+)-2-Butanol, 98+%, Thermo Scientific Chemicals
(S)-(+)-2-Butanol, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(S)-(+)-2-Butanol, 98+%, Thermo Scientific Chemicals
CAS: 4221-99-2 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFL14164.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS92-44-4
IUPAC Namenaphthalene-2,3-diol
Molecular FormulaC10H8O2
InChI KeyJRNGUTKWMSBIBF-UHFFFAOYSA-N
SMILESOC1=C(O)C=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Infrared spectrumConforms
Melting point161°C to 165°C
Appearance (Color)White to beige to pink to light grey
Appearance (Form)Powder or crystals
For synthesis of optically active products