Perfluorododecanoic acid, 96%, Thermo Scientific Chemicals
Perfluorododecanoic acid, 96%, Thermo Scientific Chemicals
Perfluorododecanoic acid, 96%, Thermo Scientific Chemicals
Perfluorododecanoic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Perfluorododecanoic acid, 96%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFL14113.031 g
Catalog number ALFL14113.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPerfluorododecanoic acid
CAS307-55-1
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.