2'-Hydroxychalcone, 95%, Thermo Scientific Chemicals
2'-Hydroxychalcone, 95%, Thermo Scientific Chemicals
2'-Hydroxychalcone, 95%, Thermo Scientific Chemicals
2'-Hydroxychalcone, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2'-Hydroxychalcone, 95%, Thermo Scientific Chemicals
CAS: 1214-47-7 | C15H12O2 | 224.259 g/mol
Have Questions?
Catalog NumberQuantity
ALFL13937.065 g
Catalog number ALFL13937.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7697-27-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormPowder
Assay (HPLC)>96.0%