Tetrahydrothiophene, 98%, Thermo Scientific Chemicals
Tetrahydrothiophene, 98%, Thermo Scientific Chemicals
Tetrahydrothiophene, 98%, Thermo Scientific Chemicals
Tetrahydrothiophene, 98%, Thermo Scientific Chemicals
Tetrahydrothiophene, 98%, Thermo Scientific Chemicals
CAS: 110-01-0 | C4H8S | 88.17 g/mol
Have Questions?
Catalog NumberQuantity
ALFL13908.22100 g
Catalog number ALFL13908.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS7447-40-7
IUPAC Namepotassium chloride
Molecular FormulaClK
InChI KeyWCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES[Cl-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric99.0 to 100.5 % (on dried substance)
Aluminium (Al)=<1 ppm
Calcium (Ca)=<50 ppm
Sodium (Na)=<700 ppm
Heavy metals (as Pb)=<5 ppm
View more
Tetrahydrothiophene is used as pharmaceutical and agrochemical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetrahydrothiophene is used as pharmaceutical and agrochemical intermediates.
Solubility
Immiscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. S. Kolboe. Catalytic hydrodesulfurization of thiophene. VII. Comparison between thiophene, tetrahydrothiophene, and n-butanethiol. Canadian Journal of Chemistry. 1969, 47, (2), 352-355.
  2. J.F. Lang; R.I. Masel. The adsorption of thiophene and tetrahydrothiophene on several faces of platinum. Surface Science. 1987, 183, (1-2), 44-66.