Isophorone diisocyanate, mixture of isomers, 98%, Thermo Scientific Chemicals
Isophorone diisocyanate, mixture of isomers, 98%, Thermo Scientific Chemicals
Isophorone diisocyanate, mixture of isomers, 98%, Thermo Scientific Chemicals
Isophorone diisocyanate, mixture of isomers, 98%, Thermo Scientific Chemicals
Isophorone diisocyanate, mixture of isomers, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isophorone diisocyanate, mixture of isomers, 98%, Thermo Scientific Chemicals
CAS: 4098-71-9 | C12H18N2O2 | 222.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
Catalog number ALFL13759.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS10297-06-0
IUPAC Name6-chlorohex-1-yne
Molecular FormulaC6H9Cl
InChI KeyZUKOCGMVJUXIJA-UHFFFAOYSA-N
SMILESClCCCCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Infrared spectrumConforms
Refractive index1.4490 to 1.4510 (20°C, 589 nm)
GC>=96.0 %
Appearance (Form)Liquid
Isophorone diisocyanate is useful for the preparation of polyurethane plastics. It is also used to make polyurethane coatings and in enamel coatings, which are resistant to abrasion and degradation from ultraviolet light. Further, it is used as coatings for automotive, flooring and roofing as well as paint applied to aircraft.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isophorone diisocyanate is useful for the preparation of polyurethane plastics. It is also used to make polyurethane coatings and in enamel coatings, which are resistant to abrasion and degradation from ultraviolet light. Further, it is used as coatings for automotive, flooring and roofing as well as paint applied to aircraft.
Solubility
Miscible with esters, ketones, ethers, and aromatic and aliphatic hydrocarbons.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and alcohols.
RUO – Research Use Only
General References:
  1. Vincent, B. J.; Natarajan, B. Kinetics of thermal degradation of water borne polyurethane dispersion containing polycaprolactone with either isophorone diisocyanate or metatetramethyl xylene diisocyanate. J. Therm. Anal. Calorim. 2015, 119 (2), 1373-1379.
  2. Dzalmukhanova, A. S.; Lodygina, V. P.; Komratova, V. V.; Karpov, S. V.; Malkov, G. V.; Badamshina, E. R. IR Spectroscopy Method for Determining The Reactivity of Isocyanate Groups in Isophorone Diisocyanate Reactions. J. Appl. Spectrosc. 2015, 82 (1), 145-148.