Phosphorus tribromide, 98%, Thermo Scientific Chemicals
Phosphorus tribromide, 98%, Thermo Scientific Chemicals
Actual product may vary
Thermo Scientific Chemicals
Phosphorus tribromide, 98%, Thermo Scientific Chemicals
CAS: 7789-60-8 | Br{3}P
Have Questions?
Catalog NumberQuantity
ALFL13638.22100 g
Catalog number ALFL13638.22
Price (MYR)
516.00
Add to cart
Quantity:
100 g
Price (MYR)
516.00
Add to cart
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point175°C
Melting Point-40°C
CAS7789-60-8
Physical FormLiquid
Quantity100 g
Molecular FormulaBr{3}P
SynonymPhosphorus(III) bromide
Percent Purity98%
Unit Size