2-(Dimethylamino)ethyl acrylate, 98%, stab. with ca 0.1% 4-methoxyphenol, Thermo Scientific Chemicals
2-(Dimethylamino)ethyl acrylate, 98%, stab. with ca 0.1% 4-methoxyphenol, Thermo Scientific Chemicals
2-(Dimethylamino)ethyl acrylate, 98%, stab. with ca 0.1% 4-methoxyphenol, Thermo Scientific Chemicals
2-(Dimethylamino)ethyl acrylate, 98%, stab. with ca 0.1% 4-methoxyphenol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-(Dimethylamino)ethyl acrylate, 98%, stab. with ca 0.1% 4-methoxyphenol, Thermo Scientific Chemicals
CAS: 2439-35-2 | C7H13NO2 | 143.19 g/mol
Have Questions?
Catalog NumberQuantity
ALFL13506.AC25 mL
Catalog number ALFL13506.AC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Request bulk or custom format
Specifications
Chemical Name or Material2-(Dimethylamino)ethyl acrylate
Name Notestabilized with ≈0.1% 4-methoxyphenol
CAS2439-35-2
Health Hazard 1H227-H302-H310+H330-H314-H317-H335
Health Hazard 2GHS H Statement
H311-H330-H314-H318-H227-H302-H317
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
May cause an allergic skin reaction.
View more
2-(Dimethylamino)ethyl acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, as a catalytic agent, as a petrochemical additive and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Dimethylamino)ethyl acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, as a catalytic agent, as a petrochemical additive and agrochemicals.
Solubility
Soluble in water (1.000 g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Sensitive to heat, light and moisture.
RUO – Research Use Only
General References:
  1. Kejian Bian.; Michael F. Cunningham. Nitroxide-mediated radical polymerization of 2-(dimethylamino)ethyl acrylate and its sequential block copolymerization with styrene and N-butyl acrylate. Journal of Polymer Science Part A: Polymer Chemistry. 2006, 44 (1), 414-426.
  2. M.S. Aziz.; Y.A. Aggourb. Electrical conduction of polymer complexes of 2-(dimethylamino)ethyl acrylate with cobalt chloride. Polymer Testing. 1999, 18 (7), 511-521.