N-tert-Butyl-2-benzothiazolesulfenamide, 97%, Thermo Scientific Chemicals
N-tert-Butyl-2-benzothiazolesulfenamide, 97%, Thermo Scientific Chemicals
N-tert-Butyl-2-benzothiazolesulfenamide, 97%, Thermo Scientific Chemicals
N-tert-Butyl-2-benzothiazolesulfenamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-tert-Butyl-2-benzothiazolesulfenamide, 97%, Thermo Scientific Chemicals
CAS: 95-31-8 | C11H14N2S2 | 238.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number ALFL13356.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN-tert-Butyl-2-benzothiazolesulfenamide
CAS95-31-8
Health Hazard 1H317
Health Hazard 2GHS H Statement
H317
May cause an allergic skin reaction.
Health Hazard 3P261-P272-P280g-P302+P352-P333+P313-P363-P501c