Tri(2-furyl)phosphine, 97%, Thermo Scientific Chemicals
Tri(2-furyl)phosphine, 97%, Thermo Scientific Chemicals
Tri(2-furyl)phosphine, 97%, Thermo Scientific Chemicals
Tri(2-furyl)phosphine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tri(2-furyl)phosphine, 97%, Thermo Scientific Chemicals
CAS: 5518-52-5 | C12H9O3P | 232.18 g/mol
Have Questions?
Catalog NumberQuantity
ALFL13329.031 g
Catalog number ALFL13329.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS13453-78-6
IUPAC Namelithium(1+) trihydrate perchlorate
Molecular FormulaClH6LiO7
InChI KeyJAWYRNYHJJDXHX-UHFFFAOYSA-M
SMILES[Li+].O.O.O.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy Metals(ICP-OES) 5 ppm
Impurity contentCl 0.003%
Impurity contentSO₄ 0.001%
pH6.0-7.5
Assay (unspecified)63.0-68.0% LiClO_4