Perchloric acid, 70% aq. soln., Thermo Scientific Chemicals
Perchloric acid, 70% aq. soln., Thermo Scientific Chemicals
Perchloric acid, 70% aq. soln., Thermo Scientific Chemicals
Perchloric acid, 70% aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Perchloric acid, 70% aq. soln., Thermo Scientific Chemicals
CAS: 7601-90-3 | HClO{4}
Have Questions?
Catalog NumberQuantity
ALFL13298.AP500 mL
Catalog number ALFL13298.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS121-00-6
IUPAC Name2-tert-butyl-4-methoxyphenol
Molecular FormulaC11H16O2
InChI KeyMRBKEAMVRSLQPH-UHFFFAOYSA-N
SMILESCOC1=CC=C(O)C(=C1)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder or waxy solid
Infrared spectrumConforms
Melting point57°C to 67°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.