Methanol, 99%, Thermo Scientific Chemicals
Methanol, 99%, Thermo Scientific Chemicals
Methanol, 99%, Thermo Scientific Chemicals
Methanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methanol, 99%, Thermo Scientific Chemicals
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1000 mL
2500 mL
Catalog number ALFL13255.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS501-98-4
IUPAC Name(2E)-3-(4-hydroxyphenyl)prop-2-enoic acid
Molecular FormulaC9H8O3
InChI KeyNGSWKAQJJWESNS-ZZXKWVIFSA-N
SMILESOC(=O)C=CC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Appearance (Color)White to cream to pale brown to green
FormPowder
Assay (HPLC)≥97.5%
Methanol is useful in the synthesis of nucleic acids and peptides. It is used as an antifreeze, solvent, fuel, and as a denaturant for ethanol. It is also used for producing biodiesel. It is used for the synthesis of formaldehyde and acetic acid. It is also useful to make chemicals, to remove water from automotive and aviation fuels, as a solvent for paints and plastics, and as an ingredient in a wide variety of products. It plays an important role in the measurement of methyl ester content of pectin. Further, it is widely used as a solvent in synthetic organic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methanol is useful in the synthesis of nucleic acids and peptides. It is used as an antifreeze, solvent, fuel, and as a denaturant for ethanol. It is also used for producing biodiesel. It is used for the synthesis of formaldehyde and acetic acid. It is also useful to make chemicals, to remove water from automotive and aviation fuels, as a solvent for paints and plastics, and as an ingredient in a wide variety of products. It plays an important role in the measurement of methyl ester content of pectin. Further, it is widely used as a solvent in synthetic organic chemistry.
Solubility
Miscible with water and most of the organic solvents including ethyl acetate, toluene, dichloromethane, chloroform, tetrahydrofuran, and acetone.
Notes
Hygroscopic. Keep the material away from heat, sparks, flame and other sources of ignition.
RUO – Research Use Only
General References:
  1. Klhar, B.; Gimenez, S.; Zandi, O.; Santiago, F. F.; Hamann, T. Competitive Photoelectrochemical Methanol and Water Oxidation with Hematite Electrodes. Appl. Mater. Interfaces 2015, 7 (14), 7653-7660.
  2. Icardi, U. A.; Specchia, S.; Fontana, G. J. R.; Saracco, G.; Specchia, V. Compact direct methanol fuel cells for portable application. J. Power Sources 2008, 176 (2), 460-467.