Methanol, 99%, Thermo Scientific Chemicals
Methanol, 99%, Thermo Scientific Chemicals
Methanol, 99%, Thermo Scientific Chemicals
Methanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methanol, 99%, Thermo Scientific Chemicals
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 mL
500 mL
1000 mL
Catalog number ALFL13255.0F
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2500 mL
Request bulk or custom format
Chemical Identifiers
CAS26555-40-8
IUPAC Name(chlorosulfanyl)(methoxy)methanone
Molecular FormulaC2H3ClO2S
InChI KeyTXJXPZVVSLAQOQ-UHFFFAOYSA-N
SMILESCOC(=O)SCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear bright yellow
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4810 to 1.4830 (20°C, 589 nm)
Methanol is useful in the synthesis of nucleic acids and peptides. It is used as an antifreeze, solvent, fuel, and as a denaturant for ethanol. It is also used for producing biodiesel. It is used for the synthesis of formaldehyde and acetic acid. It is also useful to make chemicals, to remove water from automotive and aviation fuels, as a solvent for paints and plastics, and as an ingredient in a wide variety of products. It plays an important role in the measurement of methyl ester content of pectin. Further, it is widely used as a solvent in synthetic organic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methanol is useful in the synthesis of nucleic acids and peptides. It is used as an antifreeze, solvent, fuel, and as a denaturant for ethanol. It is also used for producing biodiesel. It is used for the synthesis of formaldehyde and acetic acid. It is also useful to make chemicals, to remove water from automotive and aviation fuels, as a solvent for paints and plastics, and as an ingredient in a wide variety of products. It plays an important role in the measurement of methyl ester content of pectin. Further, it is widely used as a solvent in synthetic organic chemistry.
Solubility
Miscible with water and most of the organic solvents including ethyl acetate, toluene, dichloromethane, chloroform, tetrahydrofuran, and acetone.
Notes
Hygroscopic. Keep the material away from heat, sparks, flame and other sources of ignition.
RUO – Research Use Only
General References:
  1. Klhar, B.; Gimenez, S.; Zandi, O.; Santiago, F. F.; Hamann, T. Competitive Photoelectrochemical Methanol and Water Oxidation with Hematite Electrodes. Appl. Mater. Interfaces 2015, 7 (14), 7653-7660.
  2. Icardi, U. A.; Specchia, S.; Fontana, G. J. R.; Saracco, G.; Specchia, V. Compact direct methanol fuel cells for portable application. J. Power Sources 2008, 176 (2), 460-467.