Hydrochloric acid, 36% w/w aq. soln., Thermo Scientific Chemicals
Hydrochloric acid, 36% w/w aq. soln., Thermo Scientific Chemicals
Hydrochloric acid, 36% w/w aq. soln., Thermo Scientific Chemicals
Hydrochloric acid, 36% w/w aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrochloric acid, 36% w/w aq. soln., Thermo Scientific Chemicals
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 mL
1000 mL
Catalog number ALFL13091.0F
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2500 mL
Request bulk or custom format
Chemical Identifiers
CAS153-98-0
IUPAC Namehydrogen 3-(2-aminoethyl)-1H-indol-5-ol chloride
Molecular FormulaC10H13ClN2O
InChI KeyMDIGAZPGKJFIAH-UHFFFAOYSA-N
SMILES[H+].[Cl-].NCCC1=CNC2=CC=C(O)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale grey or pale brown
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥97.5 to ≤102.5% (Dry wt. basis, material sourced in UK)
Assay from Supplier's CofA≥97.5% (HPLC)
CommentMaterial sourced in UK or US
View more
Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing.
Solubility
Completely miscible with water.
Notes
Store in cool dry place, away from acids and alkalis.
RUO – Research Use Only
General References:
  1. Jay A. Young. Hydrochloric Acid (approx 36%) Chem. Educ. 2001, 78 (7), 873.
  2. Cui, C.; Hu, Q.; Ren, J.; Zhao, H.; You, L.; Zhao, M. Effect of the Structural Features of Hydrochloric Acid-Deamidated Wheat Gluten on Its Susceptibility to Enzymatic Hydrolysis. Agric. Food Chem. 2013, 61 (24), 5706-5714.