3-Cyanophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
3-Cyanophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
3-Cyanophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
3-Cyanophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Cyanophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
CAS: 3125-78-8 | C8H4N2S | 160.19 g/mol
Have Questions?
Catalog NumberQuantity
ALFL13045.065 g
Catalog number ALFL13045.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Cyanophenyl isothiocyanate
CAS3125-78-8
Health Hazard 1H302+H312+H332-H314-H335
Health Hazard 2GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c