Palmitic acid hydrazide, 98+%, Thermo Scientific Chemicals
Palmitic acid hydrazide, 98+%, Thermo Scientific Chemicals
Palmitic acid hydrazide, 98+%, Thermo Scientific Chemicals
Palmitic acid hydrazide, 98+%, Thermo Scientific Chemicals
Palmitic acid hydrazide, 98+%, Thermo Scientific Chemicals
CAS: 2619-88-7 | C16H34N2O | 270.461 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFL13013.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS521-31-3
IUPAC Name5-amino-1,2,3,4-tetrahydrophthalazine-1,4-dione
Molecular FormulaC8H7N3O2
InChI KeyHWYHZTIRURJOHG-UHFFFAOYSA-N
SMILESNC1=C2C(=O)NNC(=O)C2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration excess NaOH>=97.5 %
Solubility(2 % in 1 N NaOH) Clear yellow to yellow-green
Appearance (Color)Yellow to yellow-green to beige
Appearance (Form)Powder and/or chunks