4,4'-Methylenebis(N-phenylmaleimide), 95%, Thermo Scientific Chemicals
4,4'-Methylenebis(N-phenylmaleimide), 95%, Thermo Scientific Chemicals
4,4'-Methylenebis(N-phenylmaleimide), 95%, Thermo Scientific Chemicals
4,4'-Methylenebis(N-phenylmaleimide), 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4,4'-Methylenebis(N-phenylmaleimide), 95%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFL12862.1425 g
Catalog number ALFL12862.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS123-86-4
IUPAC Namebutyl acetate
Molecular FormulaC6H12O2
InChI KeyDKPFZGUDAPQIHT-UHFFFAOYSA-N
SMILESCCCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Water=<0.005 %
Infrared spectrumConforms
GC>=99.0 %
Appearance (Color)Clear colorless
4,4'-Methylenebis(N-phenylmaleimide) was used to cure glycidyl ester of eleostearic acid (GEEA).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,4′-Methylenebis(N-phenylmaleimide) was used to cure glycidyl ester of eleostearic acid (GEEA).
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Kun Huang; Zengshe Liu; Jinwen Zhang; Shouhai Li; Mei Li; Jianling Xia; Yonghong Zhou. Epoxy monomers derived from tung oil fatty acids and its regulable thermosets cured in two synergistic ways. Biomacromolecules. 2014, 15,(3), 837-843.
  2. Min Ae Han; Seon Min Woo; Kyoung-jin Min; Shin Kim; Jong-Wook Park; Dong Eun Kim; Sang Hyun Kim; Yung Hyun Choi; Taeg Kyu Kwon. 6-Shogaol enhances renal carcinoma Caki cells to TRAIL-induced apoptosis through reactive oxygen species-mediated cytochrome c release and down-regulation of c-FLIP(L) expression. Chemico-Biological Interactions. 2015, 228,69-78.