3-Hydroxy-2,2-dimethylpropionic acid, 97+%, Thermo Scientific Chemicals
3-Hydroxy-2,2-dimethylpropionic acid, 97+%, Thermo Scientific Chemicals
3-Hydroxy-2,2-dimethylpropionic acid, 97+%, Thermo Scientific Chemicals
3-Hydroxy-2,2-dimethylpropionic acid, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Hydroxy-2,2-dimethylpropionic acid, 97+%, Thermo Scientific Chemicals
CAS: 4835-90-9 | C5H10O3 | 118.132 g/mol
Have Questions?
Catalog NumberQuantity
ALFL12858.1850 g
Catalog number ALFL12858.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS141-78-6
IUPAC Nameethyl acetate
Molecular FormulaC4H8O2
InChI KeyXEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILESCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Assay (GC)≥99.5% (corrected for water)
Free acid (titration)≤0.0009meq/g
Impurity contentSubstances darkened by sulfuric acid P.T.
Water Content (Karl Fischer Titration)≤0.2%
View more
3-Hydroxy-2,2-dimethylpropionic acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Hydroxy-2,2-dimethylpropionic acid is used as pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Colleen F.Bleczinski; Clemens Richert. Solid-phase synthesis of cyclic peptide-DNA hybrids. Org. Lett. 2000, 2,(12), 1697-1700.
  2. E.Koivusalmi; E.Haatainen; K.Nikkila. RP-HPLC analysis of the compounds in the synthesis mixtures of 2, 2-dimethyl-3-hydroxypropionaldehyde by two methods combined to one instrument by a column switching technique. Journal of Liquid Chromatography & Related Technologies. 2000, 23,(16), 2447-2458.