Sodium hydrogen L-tartrate, anhydrous, 98%, Thermo Scientific Chemicals
Sodium hydrogen L-tartrate, anhydrous, 98%, Thermo Scientific Chemicals
Sodium hydrogen L-tartrate, anhydrous, 98%, Thermo Scientific Chemicals
Sodium hydrogen L-tartrate, anhydrous, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium hydrogen L-tartrate, anhydrous, 98%, Thermo Scientific Chemicals
CAS: 526-94-3 | C4H5NaO6 | 172.07 g/mol
Have Questions?
Catalog NumberQuantity
ALFL12469.22100 g
Catalog number ALFL12469.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS575-38-2
IUPAC Namenaphthalene-1,7-diol
Molecular FormulaC10H8O2
InChI KeyZUVBIBLYOCVYJU-UHFFFAOYSA-N
SMILESOC1=CC=C2C=CC=C(O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)white to yellow, grey or dark brown
FormPowder
Assay (GC)> 96.0%
Proton NMRConforms to structure
Sodium hydrogen L-tartrate is used for detecting potassium and in nutrient media. It also used as a food additive and an acidity regulator.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium hydrogen L-tartrate is used for detecting potassium and in nutrient media. It also used as a food additive and an acidity regulator.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Charoen-In, U.; Ritjareonwattu, S.; Pairwatthanaphaisan, P.; Manyum, P. Crystal Growth, Structural, Thermal, Optical, Dielectric, and Ferroelectric Characterization of Sodium Hydrogen L-Tartrate Single Crystals. Integr. Ferroelectr. 2015, 165 (1), 1-10.
  2. Bartek, L.; Klusoň, P.; Červený, L. In situ chiral modification of nickel catalysts for enantioselective hydrogenation of methyl acetoacetate. Collect. Czech. Chem. Commun. 2005, 70 (10), 1642-1652.