5-Amino-3,4-dimethylisoxazole, 97%, Thermo Scientific Chemicals
5-Amino-3,4-dimethylisoxazole, 97%, Thermo Scientific Chemicals
5-Amino-3,4-dimethylisoxazole, 97%, Thermo Scientific Chemicals
5-Amino-3,4-dimethylisoxazole, 97%, Thermo Scientific Chemicals
5-Amino-3,4-dimethylisoxazole, 97%, Thermo Scientific Chemicals
CAS: 19947-75-2 | C5H8N2O | 112.132 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number ALFL12191.04
Price (MYR)
124.17
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (MYR)
124.17
Add to cart
Specifications
Chemical Name or Material5-Amino-3,4-dimethylisoxazole
CAS19947-75-2
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Melting Point118°C to 120°C