Styrene-4-sulfonic acid sodium salt hydrate, tech. 90%, Thermo Scientific Chemicals
Styrene-4-sulfonic acid sodium salt hydrate, tech. 90%, Thermo Scientific Chemicals
Styrene-4-sulfonic acid sodium salt hydrate, tech. 90%, Thermo Scientific Chemicals
Styrene-4-sulfonic acid sodium salt hydrate, tech. 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Styrene-4-sulfonic acid sodium salt hydrate, tech. 90%, Thermo Scientific Chemicals
CAS: 2695-37-6 | C8H7NaO3S | 206.19 g/mol
Have Questions?
Catalog NumberQuantity
ALFL11685.1425 g
Catalog number ALFL11685.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS821-55-6
IUPAC Namenonan-2-one
Molecular FormulaC9H18O
InChI KeyVKCYHJWLYTUGCC-UHFFFAOYSA-N
SMILESCCCCCCCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Refractive Index1.4195-1.4235 @ 20?C
Appearance (Color)Clear colorless