3-Methyl-1-pentene, 98%, Thermo Scientific Chemicals
3-Methyl-1-pentene, 98%, Thermo Scientific Chemicals
3-Methyl-1-pentene, 98%, Thermo Scientific Chemicals
3-Methyl-1-pentene, 98%, Thermo Scientific Chemicals
3-Methyl-1-pentene, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFL11525.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS149-73-5
IUPAC Nametrimethoxymethane
Molecular FormulaC4H10O3
InChI KeyPYOKUURKVVELLB-UHFFFAOYSA-N
SMILESCOC(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.3780 to 1.3800 (20°C, 589 nm)
Appearance (Color)Clear colorless
GC>=98.5 %